Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxypyridine-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog No. p-4228778
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 312300-42-8
Molecular Formula C6H6ClNO3S
Molecular Weight (g/mol) 207.63
MDL Number MFCD06739108
InChI Key OMLIVJOTRYWHNY-UHFFFAOYSA-N
Synonym 6-methoxy-pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-6-methoxypyridine, 6-methoxypyridin-3-ylsulfonyl chloride, 2-methoxypyridine-5-sulfonylchloride, 6-methoxy-3-pyridinesulfonyl chloride, 3-pyridinesulfonylchloride, 6-methoxy, 6-methoxypyridine-3-sulphonyl chloride, 5-chlorosulfonyl-2-methoxypyridine, 6-methoxypyridine-3-yl sulfonyl chloride
PubChem CID 22344709
IUPAC Name 6-methoxypyridine-3-sulfonyl chloride
SMILES COC1=NC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 42.0°C to 45.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 207.64
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 6-Methoxypyridine-3-sulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.