Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitrofluorescein (isomer II) 98.0+%, TCI America™
SDP

Catalog No. p-7133812 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 27402-68-2
Molecular Formula C20H11NO7
Molecular Weight (g/mol) 377.308
MDL Number MFCD00059694
InChI Key GYWGQAIDMYGOLU-UHFFFAOYSA-N
PubChem CID 117391
IUPAC Name 3',6'-dihydroxy-5-nitrospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OC2=O

Specifications

Quantity 1 g
Synonym 6-Nitro-3′C,6′C-dihydroxyspiro[isobenzofuran-1(3H),9′C-(9H)xanthen]-3-one
Formula Weight 377.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitrofluorescein (isomer II)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.