Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitroindazole 98.0+%, TCI America™
SDP

Catalog No. p-7133790 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 7597-18-4
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00005695
InChI Key ORZRMRUXSPNQQL-UHFFFAOYSA-N
Synonym 6-nitroindazole, 6-nitro-2h-indazole, 6-nitroisoindazole, 1h-indazole, 6-nitro, 2h-indazole, 6-nitro, indazole,6-nitro, ccris 3263, chembl54277, 6ni, 6-nitro-indazol
PubChem CID 24239
IUPAC Name 6-nitro-1H-indazole
SMILES C1=CC2=C(C=C1[N+](=O)[O-])NN=C2

Specifications

Melting Point 182°C
Quantity 25 g
Formula Weight 163.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitroindazole
RTECSNumber NK7962100
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.