Learn More
Cayman Chemical 6-Shogaol, 555-66-8, 1 mg
SDP
Supplier:  Cayman Chemical 119011
Encompass_Preferred
Molecular Formula C17H24O3, Purity ≥95%, Formula Weight 276.4, Synonyms: 1-(4-hydroxy-3-methoxyphenyl)-4-decen-3-one; A minor, bioactive component of ginger with anti-inflammatory and anticancer properties; induces apoptosis in cancer cells through the production of reactive oxygen species and the activation of caspase, impairs tubulin polymerization, downregulates NF-B signaling, inhibits the expression of inflammatory mediators, suppresses Toll-like receptor signaling, and enhances antioxidant defense mechanisms by inducing Nrf2/ARE-mediated gene expression.
Catalog No. NC1301319
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.