Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-thien-2-ylnicotinic acid, Thermo Scientific™

Catalog No. p-4391484
Click to view available options
Quantity:
250 mg
1 g
5 g

Chemical Identifiers

CAS 179408-54-9
Molecular Formula C10H7NO2S
Molecular Weight (g/mol) 205.23
MDL Number MFCD06410384
InChI Key QYGFXNUFKLRCRB-UHFFFAOYSA-N
Synonym 6-thien-2-ylnicotinic acid, 6-thiophen-2-yl nicotinic acid, 6-2-thienyl nicotinic acid, 6-thiophen-2-yl pyridine-3-carboxylic acid, 6-2-thienyl pyridine-3-carboxylic acid, acmc-209efk, 6-thien-2ylnicotinic acid, 6-thiophen-2-yl-nicotinic acid, 6-thiophen-2-ylnicotinic acid, 2-thien-2-yl-5-pyridinecarboxylic acid
PubChem CID 15411144
SMILES OC(=O)C1=CN=C(C=C1)C1=CC=CS1

Specifications

Assay Percent Range 97%
Quantity 250 mg
IUPAC Name 6-thiophen-2-ylpyridine-3-carboxylic acid
Formula Weight 205.24
Percent Purity 97%
Chemical Name or Material 6-Thien-2-ylnicotinic acid
Hazard Category GHS Signal Word: Warning
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.