Learn More
Ace Glass, Inc. Flask, freeze drying, 600mL, 144mm height, 53mm ID neck, 100mm body OD, includes snap-on rubber cap and 3/4in ODPTFE plain manifold connection adapter
Supplier: Ace Glass, Inc. 703517