Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

608 Complete Kit, Restek™
SDP

Catalog No. 06719708 Shop All Restek Corporation Products
Click to view available options
Quantity:
1 mL

Aroclor, toxaphene, and chlordane standards may exhibit lot-to-lot variation

Copmpunds:

32022: 608 Calibration Mix

Aldrin (309-00-2)
α-BHC (319-84-6)
β-BHC (319-85-7)
δ-BHC (319-86-8)
γ-BHC (Lindane) (58-89-9)
4,4'-DDD (72-54-8)
4,4'-DDE (72-55-9)
4,4'-DDT (50-29-3)
Dieldrin (60-57-1)
Endosulfan I (959-98-8)
Endosulfan II (33213-65-9)
Endosulfan sulfate (1031-07-8)
Endrin (72-20-8)
Endrin aldehyde (7421-93-4)
Heptachlor (76-44-8)
Heptachlor epoxide (isomer B) (1024-57-3)

32006: Aroclor 1016
Aroclor 1016

32007: Aroclor 1221
Aroclor 1221

32008: Aroclor 1232
Aroclor 1232

32009: Aroclor 1242
Aroclor 1242

32010: Aroclor 1248
Aroclor 1248

32011: Aroclor 1254
Aroclor 1254

32012: Aroclor 1260
Aroclor 1260

32005: Toxaphene Solutions
Toxaphene (8001-35-2)

32021: Chlordane
Chlordane

Specifications

Concentration 2,000 μg/mL in cyclohexane
CAS 8001-35-2, 110-54-3
Packaging Ampul
Quantity 1 mL
CAS Max % 99.9%
CAS Min % 0.100000
Type Multi-component Standard
UN Number UN1208
Odor Mild
Vapor Density 2.97 (Air = 1)
Melting Point -95°C
Flash Point -8°C
Specific Gravity 0.672 g/cm3 at 15°C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.