Learn More
NEOGEN Petrifilm Aerobic Count Plates, 100 plates
The Neogen Petrifilm Aerobic Count Plate is a time-saving, sample-ready plate designed to determine total aerobic bacteria populations. A built-in grid facilitates counting colonies, helping to provide fast, precise and consistent results. Each Petrifilm Aerobic Count Plate contains a water-soluble gelling agent, nutrients and indicator — all the components needed for microbial growth with no agar preparation required. The Petrifilm Aerobic Count Plate provides total aerobic count in as little as 48 hours. A red indicator dye in the plate colors all colonies red for better contrast and easier colony counting. The red colonies are also easily distinguished from opaque food particles that can cause confusion when using other plating methods. In addition, Neogen Petrifilm Aerobic Count Plates can be used to aerobically or anaerobically enumerate lactic acid bacteria in food and environmental samples.