Learn More
NEOGEN 65% ethanol, 4 gallons
Reveal Q+ for Aflatoxin is a single-step lateral flow immunochromatographic assay based on a competitive immunoassay format intended for the quantitative testing of aflatoxin in corn and corn products.