Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7,8-Dihydroxyflavone Hydrate 98.0+%, TCI America™
SDP

Catalog No. p-7127012 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 38183-03-8
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.24
MDL Number MFCD00006836
InChI Key COCYGNDCWFKTMF-UHFFFAOYSA-N
Synonym 7,8-dihydroxyflavone, 7,8-dihydroxy-2-phenyl-4h-chromen-4-one, 7,8-dihydroxy-flavone, 4h-1-benzopyran-4-one, 7,8-dihydroxy-2-phenyl, 7,8-dhf, 7,8-dihydroxy-2-phenyl-4-benzopyrone, 7,8-dihydroxy-2-phenyl-chromen-4-one, 7,8-dihydroxyflavone hydrate, chembl75267, 7,8-dihydroxy-2-phenyl-4h-1-benzopyran-4-one
PubChem CID 1880
IUPAC Name 7,8-dihydroxy-2-phenyl-4H-chromen-4-one
SMILES OC1=CC=C2C(=O)C=C(OC2=C1O)C1=CC=CC=C1

Specifications

Melting Point 245°C
Quantity 1 g
Formula Weight 254.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7,8-Dihydroxyflavone Hydrate
RTECSNumber DJ3004000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.