Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Amino-3,4-dihydro-1(2H)-naphthalenone, 97%, Thermo Scientific Chemicals

Catalog No. p-4228309
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22009-40-1
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD07369817
InChI Key IDNLVJHOEZJNHW-UHFFFAOYSA-N
Synonym 7-amino-3,4-dihydronaphthalen-1 2h-one, 7-amino-1-tetralone, 7-amino-alpha-tetralone, 7-amino-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 7-amino-3,4-dihydro, 7-amino-3,4-dihydronaphthalen-2 1h-one, 7-amino-1,2,3,4-tetrahydronaphthalen-1-one, 7-amino-2,3,4-trihydronaphthalen-1-one, 7-amino-1-oxo-1,2,3,4-tetrahydronaphthalene, 7-aminotetralone
PubChem CID 327177
IUPAC Name 7-amino-3,4-dihydro-2H-naphthalen-1-one
SMILES NC1=CC=C2CCCC(=O)C2=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 138°C to 140°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 161.2
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 7-Amino-3, 4-dihydro-1(2H)-naphthalenone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.