Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-(Ethylamino)-4,6-dimethylcoumarin 98.0+%, TCI America™
SDP

Catalog No. p-7129349 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 26078-25-1
Molecular Formula C13H15NO2
Molecular Weight (g/mol) 217.268
MDL Number MFCD00006860
InChI Key QZXAEJGHNXJTSE-UHFFFAOYSA-N
Synonym coumarin 2, 4,6-dimethyl-7-ethylamino coumarin, 7-ethylamino-4,6-dimethyl coumarin, 7-ethylamino-4,6-dimethyl-2h-chromen-2-one, 7-ethylamino-4,6-dimethyl-chromen-2-one, 4,6-dimethyl-7-ethylaminocoumarin, coumarin, 7-ethylamino-4,6-dimethyl, 2h-1-benzopyran-2-one, 7-ethylamino-4,6-dimethyl, coumarin 450, ccris 4958
PubChem CID 96929
IUPAC Name 7-(ethylamino)-4,6-dimethylchromen-2-one
SMILES CCNC1=C(C=C2C(=CC(=O)OC2=C1)C)C

Specifications

Melting Point 167°C
Quantity 200 mg
Formula Weight 217.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Ethylamino)-4,6-dimethylcoumarin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.