Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Fluorobenzo-2-oxa-1,3-diazole-4-sulfonic acid ammonium salt, MP Biomedicals™

Catalog No. p-4774482 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5 mg
10 mg
25 mg

Sensitive and specific fluorescent labeling reagent for low molecular weight compounds and for macromolecules containing thiol groups

Chemical Identifiers

CAS 84806-27-9
Molecular Formula C6H6FN3O4S
Molecular Weight (g/mol) 235.189
MDL Number MFCD02682109
InChI Key JXLHNMVSKXFWAO-UHFFFAOYSA-N
Synonym sbd-f, 7-fluorobenzofurazan-4-sulfonic acid ammonium salt, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate, azanium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonate, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonic acid, azamethane, 2,1,3-benzoxadiazole-4-sulfonic acid, 7-fluoro-, ammonium salt 1:1
PubChem CID 2737780
IUPAC Name azanium;7-fluoro-2,1,3-benzoxadiazole-4-sulfonate
SMILES C1=C(C2=NON=C2C(=C1)S(=O)(=O)[O-])F.[NH4+]

Specifications

Quantity 5 mg
Formula Weight 235.2
Physical Form Powder
Chemical Name or Material 7-Fluorobenzo-2-oxa-1, 3-diazole-4-sulfonic acid ammonium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.