Learn More
7-Hydroxycoumarin-3-carboxylic Acid 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Specifications
Melting Point
263°C
Color
Yellow
Quantity
1 g
Formula Weight
206.15
Percent Purity
≥98.0% (HPLC)
Physical Form
Crystalline Powder
Chemical Name or Material
7-Hydroxycoumarin-3-carboxylic Acid
TSCA
No
Recommended Storage
Freezer
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.