Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
7-Hydroxycoumarin-3-carboxylic Acid 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
200 mg
1 g
Specifications
Specifications
Melting Point | 263°C |
Color | Yellow |
Quantity | 200 mg |
Formula Weight | 206.15 |
Percent Purity | ≥98.0% (HPLC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 7-Hydroxycoumarin-3-carboxylic Acid |
Safety and Handling
TSCA | No |
Recommended Storage | Freezer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction