Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Nitroindole-2-carboxylic acid, 90%, Thermo Scientific™

Catalog No. KM09181DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 6960-45-8
Molecular Formula C9H6N2O4
Molecular Weight (g/mol) 206.157
InChI Key BIUCOFQROHIAEO-UHFFFAOYSA-N
Synonym 7-nitro-1h-indole-2-carboxylic acid, 7-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 7-nitro, 7-nitroindole-2-carboxylicacid, dna base excision repair pathway inhibitor, maybridge1_004786, indole-2-carboxylic acid, 7-nitro, pubchem9857, acmc-1b8pb, 1h-indole-2-carboxylicacid, 7-nitro
PubChem CID 81409
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NC(=C2)C(=O)O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 206.16
Percent Purity 90%
Chemical Name or Material 7-Nitro-1H-indole-2-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.