Learn More
Alkali Scientific 70 um Cell Strainer, PP, Gamma Sterilized, 50/pk
Supplier: Alkali Scientific TS70
ASI™ Cell Strainers make obtaining a more uniform single cell easy. Individually packaged and gamma sterilized. ASI cell strainers are the perfect choice for your research. • Individually Wrapped Pre-sterilized • Reduced Chance of Contamination • Nylon Mesh Filtration • Fits Standard 50mL Conical Tubes • Diameter bottom: 27.05mm, Height:14.50 mm • SAL 10^-6 sterilization • QTY: 50 Units/Case
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.