Learn More
Technical Manufacturing Corpor 750 x 1500 mm CleanBench Table with Gimbal Piston vibration isolation legs, top with metric tapped holes (M6, 25mm centers)
SDP
Supplier:  Technical Manufacturing Corpor 637515M
Encompass_Preferred
750 x 1500 mm CleanBench Table with Gimbal Piston vibration isolation legs, top with metric tapped holes (M6, 25mm centers)
Catalog No. NC2068796
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.