Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromoisoquinoline 97.0+%, TCI America™
SDP

Catalog No. p-7123377 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 63927-22-0
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD04973298
InChI Key DPRIHFQFWWCIGY-UHFFFAOYSA-N
Synonym 8-bromo-isoquinoline, isoquinoline,8-bromo, isoquinoline, 8-bromo, d-lactosyl fluoride, pubchem6213, acmc-1b6s3, ksc352q0f, abbypharma ap-12-10879
PubChem CID 9859134
IUPAC Name 8-bromoisoquinoline
SMILES BrC1=C2C=NC=CC2=CC=C1

Specifications

Melting Point 78°C
Color Yellow
Quantity 1 g
Formula Weight 208.06
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Bromoisoquinoline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.