Learn More
AdipoGen 8-Hydroxyjulolidine-9-aldehyde
SDP
Supplier:  AdipoGen CDXH0021G001
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 63149-33-7. Formula C13H15NO2. MW 217.26. Synthetic. Probe that exhibits pH sensitive detection of hydrogen sulfide and silicate in aqueous solution. It displays an excellent selectivity for hydrogen sulfide and silicate over other weak acid salts. The probe detects S2− in live cells, providing a powerful method to study H2S chemistry in biological systems. Used also as an intermediate and building block to synthesize coumarins and other fluorescent compounds and small molecule inhibitors.
Catalog No. 50-256-0802