Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Methyl-1,2,3,4-tetrahydroquinoline, 97%, Thermo Scientific™

Catalog No. BTB00733DA
Click to view available options
:
Amber Glass Bottle; 1g

Specifications

CAS 52601-70-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H13N
Quantity 1g
MDL Number MFCD00218100
Synonym 8-methyl-1,2,3,4-tetrahydro-quinoline, 8-methyl-1,2,3,4-tetrahydroquinolin, 1,2,3,4-tetrahydro-8-methylquinoline
InChI Key YIIPMCFBCZKCFB-UHFFFAOYSA-N
SMILES CC1=CC=CC2=C1NCCC2
IUPAC Name 8-methyl-1,2,3,4-tetrahydroquinoline
Molecular Weight (g/mol) 147.221
PubChem CID 2774166
Formula Weight 147.22
Percent Purity 97%
Chemical Name or Material 8-Methyl-1,2,3,4-tetrahydroquinoline
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.