Learn More
AdipoGen 9 10-Anthracenediyl-bismethyl
SDP
Supplier:  AdipoGen CDXA0183G001
Encompass_Preferred
Chemical. CAS 307554-62-7. Formula C22H18O8. MW 410.37. Synthetic. Reagent for the assay of O2 with better characteristics than 9,10-anthracenediyl-bis-dipropionic acid.
Catalog No. 50-256-1809
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.