Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7134319 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 82419-35-0
Molecular Formula C13H9F2NO4
Molecular Weight (g/mol) 281.215
MDL Number MFCD00226106
InChI Key NVKWWNNJFKZNJO-UHFFFAOYSA-N
Synonym 9,10-difluoro-2,3-dihydro-3-methyl-7-oxo-7h-pyrido 1,2,3-de-1,4-benzoxazine-6-carboxylic acid, ofloxacin q acid, oxygen-fluorine acid, 9,10-difluoro-3-methyl-7-oxo-2,3-dihydro-7h-1,4 oxazino 2,3,4-ij quinoline-6-carboxylic acid, s---9,10-difluoro-2,3-dihydro-3-methyl-7-oxo-7h-pyrido-1,2,3-de-1,4-benzoxazine-6-carboxylic acid, 9,10-difluoro-2,3-dihydro-3-methyl-7-oxo-7h-pyrido 1,2,3-de-1,4-benzoxazine-6-carboxyllic acid, 9,10-difluoro-3-methyl-7-oxo-2,3-dihydro-7h-pyrido 1,2,3-de 1,4 benzoxazine-6-carboxylic acid, levofloxacin related compound b, ofloxacin q acid, +/-, 6,7-difluoro-2-methyl-10-oxo-4-oxa-1-azatricyclo 7.3.1.0?, 1 3 trideca-5 13 ,6,8,11-tetraene-11-carboxylic acid
PubChem CID 534278
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2F)F)C(=O)O

Specifications

Melting Point 311°C
Color White-Yellow
Quantity 5 g
Formula Weight 281.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.