Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Bromo-10-(1-naphthyl)anthracene 98.0+%, TCI America™
SDP

Catalog No. p-7123546 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 400607-04-7
Molecular Formula C24H15Br
Molecular Weight (g/mol) 383.288
MDL Number MFCD11046571
InChI Key SYACRXBYRNYMLN-UHFFFAOYSA-N
Synonym 9-bromo-10-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl anthracene, 9-bromo-10-1-naphthyl anthracene, anthracene, 9-bromo-10-1-naphthalenyl, 9-bromo-10-naphthalen-1-yl-anthracene, 10-bromo-9-phthalen-1-yl anthracene, 10-bromo-9-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl-anthracene, 10-naphthalene-1-yl-9-bromo anthracene
PubChem CID 21076365
IUPAC Name 9-bromo-10-naphthalen-1-ylanthracene
SMILES C1=CC=C2C(=C1)C=CC=C2C3=C4C=CC=CC4=C(C5=CC=CC=C53)Br

Specifications

Melting Point 179°C
Color Yellow
Quantity 1 g
Formula Weight 383.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromo-10-(1-naphthyl)anthracene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.