Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Bromo-10-[4-(2-naphthyl)phenyl]anthracene 98.0+%, TCI America™
SDP

Catalog No. p-7123626 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 866611-29-2
Molecular Formula C30H19Br
Molecular Weight (g/mol) 459.386
MDL Number MFCD20486476
InChI Key PIGVQJMWWHSAEZ-UHFFFAOYSA-N
PubChem CID 23148608
IUPAC Name 9-bromo-10-(4-naphthalen-2-ylphenyl)anthracene
SMILES C1=CC=C2C=C(C=CC2=C1)C3=CC=C(C=C3)C4=C5C=CC=CC5=C(C6=CC=CC=C64)Br

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 459.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromo-10-[4-(2-naphthyl)phenyl]anthracene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.