Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenylmethyl pentafluorophenyl carbonate, 98%, Thermo Scientific™

Catalog No. AC303080050
Click to view available options
Quantity:

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS

88744-04-1

Molecular Formula

C21H11F5O3

Molecular Weight (g/mol)

406.308

MDL Number

MFCD00013261

InChI Key

CBBKZVZOEBSFQX-UHFFFAOYSA-N

Synonym

9-fluorenylmethyl pentafluorophenyl carbonate, fmoc-opfp, 9h-fluoren-9-yl methyl perfluorophenyl carbonate, 9h-fluoren-9-ylmethyl 2,3,4,5,6-pentafluorophenyl carbonate, 9h-fluoren-9-ylmethyl pentafluorophenyl carbonate, carbonic acid 9h-fluoren-9-ylmethyl pentafluorophenyl ester, carbonic acid,9h-fluoren-9-ylmethyl 2,3,4,5,6-pentafluorophenyl ester, acmc-20ecck, fmoc pentafluorophenyl ester, 9-fluorenylmethylpentafluorophenylcarbonate

PubChem CID

2733201

IUPAC Name

9H-fluoren-9-ylmethyl (2,3,4,5,6-pentafluorophenyl) carbonate

SMILES

C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)OC4=C(C(=C(C(=C4F)F)F)F)F

Specifications

CAS Min %

97.5

CAS Max %

100.0

Melting Point

83°C to 87°C

Color

White

Elemental Analysis

97.5% min. (ex C)

Infrared Spectrum

Authentic

Assay Percent Range

98%

Quantity

5 g

Formula Weight

406.31

Physical Form

Powder

Chemical Name or Material

9-Fluorenylmethyl pentafluorophenyl carbonate, 98%

Hazard Category

MOISTURE SENSITIVE

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.