Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Trifluoroacetylanthracene 98.0+%, TCI America™
SDP

Catalog No. p-7117425 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 53531-31-0
Molecular Formula C16H9F3O
Molecular Weight (g/mol) 274.242
MDL Number MFCD00001258
InChI Key MNCMBBIFTVWHIP-UHFFFAOYSA-N
Synonym 9-trifluoroacetylanthracene, 9-anthryl trifluoromethyl ketone, 1-anthracen-9-yl-2,2,2-trifluoroethanone, 9-trifluoroacetyl anthracene, 1-9-anthryl-2,2,2-trifluoroethan-1-one, 1-9-anthryl-2,2,2-trifluoroethanone, 9-anthryl, pubchem16290, acmc-209l8i, bidd:gt0598
PubChem CID 104525
IUPAC Name 1-anthracen-9-yl-2,2,2-trifluoroethanone
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2C(=O)C(F)(F)F

Specifications

Melting Point 85°C
Color Yellow
Quantity 1 g
Formula Weight 274.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Trifluoroacetylanthracene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.