Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ 968
Allosteric inhibitor of glutaminase
Specifications
Purity | >99 |
---|---|
Description | 968 |
Molecular Weight (g/mol) | 475.42 |
Product Type | 968 |
Description
Allosteric inhibitor of glutaminase. Blocks Rho-GTPase-dependent transformation of fibroblasts. Suppresses breast cancer cell growth and invasive activity in vitro. Also inhibits P-493 B lymphoma cell growth and reduces tumor size of P-493 cell xenografts in mice.Specifications
>99 | |
968 | |
475.42 | |
968 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title