Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A 205804, Tocris Bioscience™

Catalog No. p-4783697 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective inhibitor of E-selectin and ICAM-1 expression

Selective inhibitor of E-selectin and ICAM-1 expression (IC50 values are 20, 25 and > 1000 nM for TNF-α-induced E-selectin, ICAM-1 and VCAM-1 expression respectively). Effective inhibitor of cell-cell adhesion in an in vitro flow experiment.

Chemical Identifiers

CAS 251992-66-2
Molecular Formula C15H12N2OS2
Molecular Weight (g/mol) 300.394
InChI Key QQGWEXFLMJGCAL-UHFFFAOYSA-N
Synonym 4-p-tolylthio thieno 2,3-c pyridine-2-carboxamide, unii-tp7tb1sszd, tp7tb1sszd, 4-4-methylphenyl thio thieno 2,3-c pyridine-2-carboxamide, 4-p-tolylsulfanyl thieno 2,3-c pyridine-2-carboxamide, thieno 2,3-c pyridine-2-carboxamide, 4-4-methylphenyl thio, 4-4-methylphenyl thio thieno 2,3-c pyridine-2-ca, 4-p-tolylsulfanyl-thieno 2,3-c pyridine-2-carboxylic acid amide
PubChem CID 9839311
IUPAC Name 4-(4-methylphenyl)sulfanylthieno[2,3-c]pyridine-2-carboxamide
SMILES CC1=CC=C(C=C1)SC2=C3C=C(SC3=CN=C2)C(=O)N

Specifications

Quantity 10 mg
Formula Weight 300.39
Percent Purity >98%
Chemical Name or Material A 205804
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.