Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A 68930 hydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective D1-like dopamine receptor agonist (EC50 values are 2.1 and 3910 nM for D1-like and D2-like receptors respectively). Centrally active following systemic administration in vivo.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 307.78 |
Percent Purity | >98% |
Chemical Name or Material | A 68930 hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction