Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A 769662, Tocris Bioscience™

Catalog No. p-4783700 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent AMPK activator

Potent, reversible AMP-activated protein kinase (AMPK) activator (EC50 = 0.8 μM) that displays selectivity towards β1 subunit-containing heterotrimers. Inhibits fatty acid synthesis (IC50 = 3.2 μM) and decreases plasma glucose and triglyceride levels in vivo. Also inhibits proliferation of mesenchymal stem cells, and impedes reprogramming of mouse embryonic fibroblasts to iPSCs. Activates autophagy.

Chemical Identifiers

CAS 844499-71-4
Molecular Formula C20H12N2O3S
Molecular Weight (g/mol) 360.387
InChI Key CTESJDQKVOEUOY-UHFFFAOYSA-N
Synonym 4-hydroxy-3-2'-hydroxy-1,1'-biphenyl-4-yl-6-oxo-6,7-dihydrothieno 2,3-b pyridine-5-carbonitrile, unii-p68477cd2c, 6,7-dihydro-4-hydroxy-3-2'-hydroxy 1,1'-biphenyl-4-yl-6-oxo-thieno 2,3-b pyridine-5-carbonitrile, 4-hydroxy-3-2'-hydroxybiphenyl-4-yl-6-oxo-6,7-dihydrothieno 2,3-b pyridine-5-carbonitrile, 4-hydroxy-3-4-2-hydroxyphenyl phenyl-6-oxo-7h-thieno 2,3-b pyridine-5-carbonitrile, thieno 2,3-b pyridine-5-carbonitrile, 6,7-dihydro-4-hydroxy-3-2'-hydroxy 1,1'-biphenyl-4-yl-6-oxo, 4cff, 4-hydroxy-3-2'-hydroxy-1,1'-biphenyl-4-yl-6-oxo-7h-thieno 2,3-b pyridine-5-carbonitrile
PubChem CID 54708532
IUPAC Name 4-hydroxy-3-[4-(2-hydroxyphenyl)phenyl]-6-oxo-7H-thieno[2,3-b]pyridine-5-carbonitrile
SMILES C1=CC=C(C(=C1)C2=CC=C(C=C2)C3=CSC4=C3C(=C(C(=O)N4)C#N)O)O

Specifications

Quantity 10 mg
Formula Weight 360.39
Percent Purity >98%
Chemical Name or Material A 769662
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.