Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A 803467, Tocris Bioscience™

Catalog No. p-4783702 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective NaV1.8 channel blocker

Selective blocker of NaV1.8 channels (IC50 values are 8, 2450, 6740, 7340 and 7380 nM for hNaV1.8, hNaV1.3, hNaV1.7, hNaV1.5 and hNaV1.2 channels respectively). Shows no significant activity against TRPV1, P2X2/3, CaV2.2 and KCNQ2/3 channels. Antinociceptive; potently attenuates mechanical allodynia in two models of neuropathic pain following i.p. administration.

Chemical Identifiers

CAS 944261-79-4
Molecular Formula C19H16ClNO4
Molecular Weight (g/mol) 357.79
MDL Number MFCD10574689
InChI Key VHKBTPQDHDSBSP-UHFFFAOYSA-N
PubChem CID 16038374
IUPAC Name 5-(4-chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide
SMILES COC1=CC(OC)=CC(NC(=O)C2=CC=C(O2)C2=CC=C(Cl)C=C2)=C1

Specifications

Quantity 10 mg
Synonym 5-4-chlorophenyl-n-3,5-dimethoxyphenyl furan-2-carboxamide, 5-4-chlorophenyl-n-3,5-dimethoxyphenyl-2-furancarboxamide, d0ff6s, 2-furancarboxamide, 5-4-chlorophenyl-n-3,5-dimethoxyphenyl, 5-4-chlorophenyl-n-3,5-dimethoxyphenyl, 5-4-chlorophenyl-furan-2-carboxylic acid 3,5, a hplc, 5-4-chlorophenyl-n-3,5-dimethoxyphenyl-2-furamide, 2-furancarboxamide,5-4-chlorophenyl-n-3,5-dimethoxyphenyl
Formula Weight 357.79
Percent Purity >98%
Chemical Name or Material A 803467
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.