Learn More
Revvity Health Sciences Inc A1R expressing cells labeled with Terbium, 200 Tests
Supplier: Revvity Health Sciences Inc C1TT1A1
A1 receptor is one of the four adenosine receptor family (including A2A, A2B and A3 receptors). It has vital role in cardiac and renal processes. It regulates neurotransmitters in the brain, such as dopamine and glutamate. Thus, it is considered as an interesting target in treatment of ischemia-reperfusion injury, renal pathologies, and neuropathic pain. Cells expressing the A1 receptor are provided pre-labeled with Terbium, and can be used to conduct receptor binding studies on the aforementioned receptor.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.