Learn More
Teknova AAV Affinity CIP1 Solution, 1000mL, Sterile 8/UN1760/III
SDP
Supplier:  Teknova 20770
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
0.1M Phosphoric Acid adjusted to pH 2.0
Catalog No. 50-238-7467