Learn More
TARGETMOL CHEMICALS INC AB928 5MG
Supplier:  TARGETMOL CHEMICALS INC T14078
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AB928 is an orally bioavailable selective dual adenosine receptor (A2aR/A2bR) antagonist. It has immunomodulatory activity[1]. purity: 98%
Catalog No. 50-257-7389
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.