Learn More
TARGETMOL CHEMICALS INC ABEPRAZAN HYDROCHLORIDE 5MG
Supplier:  TARGETMOL CHEMICALS INC T10221
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Abeprazan hydrochloride (Fexuprazan hydrochloride) is an effective reversible potassium-competitive acid blocker with oral activity inhibiting H+ K+ -atPase by competitive binding to potassium ions without acid activation. Abeprazan hydrochloride is a proton pump inhibitor (PPI) that acts by reducing gastric acid production and is used to treat gastric acid-related disorders such as gastroesophageal reflux disease (GERD) and peptic ulcers. purity: 98%
Catalog No. 50-258-1745