Learn More
Abnova™ Hydrogen Peroxide Chemiluminescent Detection Kit
Shop All Abnova Corporation Products- For the quantitative determination of hydrogen peroxide in colorless tissue culture media and buffers
- Measurement helps to determine how oxidative stress modulates varied intracellular pathways
- All components of this kit are stable at 4°C until the expiration date
Specifications
Description
Hydrogen Peroxide Chemiluminescent Detection Kit is a higher throughput assay for the quantitative measurement of hydrogen peroxide in culture supernatants and cell lysates from any species.
Product Type
Hydrogen Peroxide Chemiluminescent Detection Kit
Sample Type
Cell Lysates, Cell Culture Supernatant
For Use With (Application)
Quantification
Kit Contents
Hydrogen peroxide standard 0.5 mL, Hydrogen peroxide trigger solution 15 mL, Hydrogen peroxide trigger solution 6 mL, Microwell 96-well
Quantity
1 Kit
Sample Volume
50 μL
Storage Requirements
Store the kit at 4°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.