Learn More
TARGETMOL CHEMICALS INC ABT-702 DIHYDROCHLORIDE 5MG
Supplier: TARGETMOL CHEMICALS INC T4668
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ABT-702 dihydrochloride is a potent adenosine kinase (AK) inhibitor. purity: 98%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.