Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AC 261066, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent RARβ2 agonist (pEC50 = 8.1). Selective over RARβ1, RARα and RARγ (pEC50 values are 6.4, 6.2 and 6.3 respectively). More potent and orally available than AC 55649 (Cat. No. 2436).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 353.41 |
Percent Purity | >98% |
Chemical Name or Material | AC 261066 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction