Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Barricade Tapes
High-visibility barricade tapes are a quick, temporary and effective way to surround or section-off hazard areas, danger zones and construction sites
$73.00
Specifications
Adhesive Type | Non-adhesive |
---|---|
Dimensions (L x W) | 1000 ft. x 3 in. (304.8 m x 7.62 cm) |
Material | HDPE |
For Use With (Application) | Crowd control |
Length (English) | 1000 ft. |
Catalog Number | Mfr. No. | Color | Displayed Text | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Displayed Text | Price | Quantity & Availability | ||||
19-804-923
|
Accuform Signs
MPT05 |
Black, Red | Danger Asbestos Hazard |
Each for $73.00
|
|
||||
19-804-925
|
Accuform Signs
MPT139 |
Black, Yellow | Police Line Do Not Cross |
Each for $73.00
|
|
||||
19-804-926
|
Accuform Signs
MPT24 |
Red, Black | Danger Keep Out |
Each for $73.00
|
|
||||
Description
- Size: 7.6cm × 304.8m (3 in. × 1000 ft.)
- Material: Polyethylene plastic
- Non-adhesive
- Self-wound on a roll
- Unspool, cut easily, and can be fastened with any method
- Clear, bold warning message that is continuously repeated the length of the roll
- Material is economical and disposable, yet can be reused
Specifications
Non-adhesive | |
HDPE | |
1000 ft. | |
3 mil | |
7.62 cm |
1000 ft. x 3 in. (304.8 m x 7.62 cm) | |
Crowd control | |
304.8 m | |
3 in. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title