Learn More
SeraCare™ ACCURUN 1™ Multi-Analyte Independent Positive Controls
Shop All SeraCare Life Sciences ProductsACCURUN 1 controls are formulated for use with in vitro diagnostic test methods for the qualitative detection of HBsAg and antibodies or antigens to multiple bloodborne viruses
- Multi-Analyte Positive Controls for Hepatitis (HBc, HCV, HBsAg), HIV (1 and 2), HTLV (I and II) and CMV
- Helps to enhance quality control and quality assurance in testing labs
- Human plasma based controls mimic human samples
- Liquid, ready-to-use - no reconstitution or special handling required
- Stable at refrigerated temperatures
- Controls are specially formulated as low level positive for most manual and automated assay platforms
- Independent controls meet compliance requirements of regulatory agencies and certifying organizations
- Extended shelf life means efficiency for labs, allowing assay performance to be monitored with same lot over an extended period of time
- SeraCare facilities are ISO 9001 and 13485 certified and comply with cGMP standards
Certifications
CE marked
Compliance
IVD, HC (22-554-500, 22-554-469, 22-554-470, 22-554-301, 21-123-102)
Specifications
Certifications/Compliance
CE, IVD, CE, HC
Content And Storage
2°C to 8°C
Format
Tube
For Use With (Equipment)
Abbott Prism™
Type
ACCURUN 1 Series 2500
Intended Use
Detection of Antibodies to HIV 1 and 2, Antibodies to HTLV I and II, Antibodies to HBcAg, Antibodies to HCV, and HBsAg
Quantity
12 x 3.5 mL
Source
Human Serum or Plasma
Stability
60 Days Open Vial
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.