Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SeraCare™ ACCURUN™ QC Hepatitis Positive Controls
Offer the ultimate in flexibility and convenience
$372.00 - $476.00
Specifications
Certifications/Compliance | IVD, EU, CE |
---|---|
Content And Storage | 2°C to 8°C |
Format | Vial |
Quantity | 1 x 5 mL |
Source | Human Serum or Plasma |
Catalog Number | Mfr. No. | Type | Intended Use | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Intended Use | Quantity | Price | Quantity & Availability | |||
22-554-173
|
SeraCare™
20150077 |
ACCURUN 125 anti-HBs Positive Control | Qualitative detection of Antibodies to Hepatitis B Surface Antigen (Anti-HBs) | 1 x 5 mL |
Pack for $372.00
|
|
|||
22-554-171
|
SeraCare™
20150074 |
ACCURUN 121 Anti-HAV IgM Positive Control | Qualitative determination of IgM Antibodies to Hepatitis A Virus (HAV IgM) | 1 x 5 mL |
Pack for $430.00
|
|
|||
22-554-165
|
SeraCare™
20150068 |
ACCURUN 113 HBc IgM Positive Control | Detection of IgM Antibodies to Hepatitis B core Antigen (HBc IgM) | 1 x 5 mL |
Pack for $476.00
|
|
|||
Description
- Manufactured from human serum or plasma
Specifications
IVD, EU, CE | |
Vial | |
Human Serum or Plasma |
2°C to 8°C | |
1 x 5 mL | |
60 Days Open Vial |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title