Learn More
Bioassay Systems EnzyFluo™ Acetaldehyde Assay Kit. For quantitatve determination of acetaldehyde and evaluations of drug effects on its metabolism.
SDP
Supplier:  Bioassay Systems EFAC100
Encompass_Preferred
For quantitatve determination of acetaldehyde and evaluations of drug effects on its metabolism. Key Features: Fast and sensitive. Linear detection range (50µL sample): 0.5 µM to 60 µM acetaldehyde in 96-well plate assay. High-throughput. The procedure involves adding a single working reagent, and reading the absorbance after 30 minutes. Room temperature assay. No 37°C heater is needed. Convenient. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: FL530/585nm. Samples: Biological samples (e.g. plasma, serum, urine, tissue and culture media) and food/beverage samples (e.g. wine, coffee, and juice). Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.5 µM.
Catalog No. 50-225-5875
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.