Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Acid, Glacial, Aldehyde Free, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500mL
2.5L
4L
Packaging:
Clear Glass Bottle
Clear Glass Bottle in Single Shipper
Clear PVC Coated Glass Bottle
Glass Bottle
PVC Coated Glass Bottle
Poly Bottle
Specifications
Specifications
Formula Weight | 60.05 |
Grade | Meets ACS reagent chemical requirements |
Packaging | Clear Glass Bottle in Single Shipper |
Specific Gravity | 1L = 1.05kg |
Quantity | 500mL |
Flash Point | 39.00°C |
Chemical Name or Material | Acetic Acid, Glacial, Aldehyde Free |
Safety and Handling
Recommended Storage | IMPORTANT: Material will freeze if stored below 17°C (63°F) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction