Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Acetic Anhydride
SCI_ED

Catalog No. p-4416520 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Lab Grade
Reagent Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate
SMILES CC(=O)OC(=O)C

Specifications

Density 1.0820g/cmsuperscript_3;
Assay Percent Range >97% w/v
Packaging Lab Grade
Linear Formula (CH3CO)2O
Quantity 100 mL
Formula Weight 102.09
Chemical Name or Material Acetic Anhydride
ShelfLife 5 years
DOTInformation DOT Class 8 (3), II, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.