Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetone (ACS AR), Macron Fine Chemicals™
Shop All Avantor Products
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
Packaging:
Glass
Poly
Poly bottle
Specifications
Specifications
Quantity | 1 L |
Packaging | Glass |
Formula Weight | 58.08 |
Grade | ACS Reagent |
Chemical Name or Material | Acetone |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction