Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile/Formic Acid, 0.1%, Mobile Phase, MilliporeSigma™

Catalog No. 50947839 Shop All MilliporeSigma Products
Click to view available options
Quantity:
4 L

Chemical Identifiers

CAS 64-18-6

Specifications

Assay Percent Range 0.095 to 0.105% (v/v)
Quantity 4 L
Flash Point 5.9°C
Packaging Glass Bottle, Pre-Packed
Residue after Evaporation 5 ppm max
Recommended Storage Store at room temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.