Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetophenone O-Benzoyloxime 98.0+%, TCI America™
SDP

Catalog No. p-7117946 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 26060-56-0
Molecular Formula C15H13NO2
Molecular Weight (g/mol) 239.274
InChI Key KLJLQTJYNGGTIU-FOWTUZBSSA-N
PubChem CID 5900887
IUPAC Name [(E)-1-phenylethylideneamino] benzoate
SMILES CC(=NOC(=O)C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 100°C
Color White
Quantity 1 g
Formula Weight 239.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Acetophenone O-Benzoyloxime
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.