Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acridine Orange hydrochloride, Tocris Bioscience™

Catalog No. 5092 Shop All R&D Systems Products
Click to view available options
Quantity:
This item is not returnable. View return policy
This item is not returnable. View return policy

Nucleic acid binding dye

Cell and organelle membrane permeable nucleic acid binding dye. Emits green fluorescence when bound to double-stranded DNA and red fluorescence when bound to RNA or single-stranded DNA. Used in cell cycle and apoptosis studies and as a lysosomal dye.

Chemical Identifiers

CAS

65-61-2

Molecular Formula

C17H20ClN3

Molecular Weight (g/mol)

301.82

MDL Number

MFCD00012660

InChI Key

ASKSWIKIIFUEOI-UHFFFAOYSA-N

Synonym

acridine orange hydrochloride, c.i. basic orange 14, rhoduline orange, acridine orange n, 3,6-bis dimethylamino acridine hydrochloride, basic orange 14, basic orange 3rn, acridine orange no, acridine orange r, rhoduline orange no

PubChem CID

517204

ChEBI

CHEBI:51739

IUPAC Name

hydrogen N1,N1,N7,N7-tetramethylacridine-1,7-diamine chloride

SMILES

[H+].[Cl-].CN(C)C1=CC=C2N=C3C=CC=C(N(C)C)C3=CC2=C1

Specifications

Color

Orange

Quantity

50 mg

Formula Weight

301.81

Percent Purity

>99%

Physical Form

Solid

Chemical Name or Material

Acridine Orange hydrochloride

Recommended Storage

Store at RT

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.