Learn More
Cytiva (Formerly Pall Lab) AcroPrep™ 384-well Filter Plates, 100 L - 10K Omega membrane, long tips, natural (10/pkg)
Supplier:  Cytiva 5077
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Ideal for Use With Low Sample Volume, High Throughput Sample Prep Applications-Plates are constructed from chemically-resistant, biologically-inert polypropylene with a clear polystyrene lid.-Robotic-friendly design has single-piece construction, designed in accordance with the ANSI/SBS X-2004 standards. Plates are stackable with and without lids.-Eliminates crosstalk between wells. Membranes are individually sealed in each well with a proprietary sealing process and each well has a fluid director.-A serialized barcode label allows the use of automated tracking systems and identifies the membrane type.-Available in a variety of membrane configurations to meet the requirements of numerous applications. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3131